Product Image
Thermo Scientific™

EZ-Link™ NHS-PEG Solid-Phase Biotinylation Kit - Mini-Spin Columns

Thermo Scientific EZ-Link NHS-PEG Solid Phase Biotinylation Kits provide an on-column approach to amine-targeted antibody biotinylation that provides greater controlRead more
Have Questions?
Catalog NumberQuantity
214508 reactions
Catalog number 21450
Price (INR)
35,503.00
Online offer
Ends: 31-Dec-2026
59,171.00
Save 23,668.00 (40%)
Each
Quantity:
8 reactions
Request bulk or custom format
Price (INR)
35,503.00
Online offer
Ends: 31-Dec-2026
59,171.00
Save 23,668.00 (40%)
Each
Thermo Scientific EZ-Link NHS-PEG Solid Phase Biotinylation Kits provide an on-column approach to amine-targeted antibody biotinylation that provides greater control over the labeling reaction and clean-up process.

Features of the EZ-Link NHS-PEG Solid-Phase Biotinylation Kit:

• Suitable for larger-scale reactions—1 to 10 mg antibody
• Fast labeling and purification—the entire procedure takes only about one hour
• Easy removal of spent and excess labeling reagent—simply wash away the reaction byproducts—no need for dialysis or gel filtration
• No dilution effects—solid-phase method allows initially dilute antibodies to be recovered in a smaller volume after labeling
• Optimized protocols—specific protocols for antibody ensure appropriate level of labeling (2 to 5 biotins per antibody molecule), minimizing possibility of inactivation caused by overlabeling
• High-performance biotin reagent—NHS-PEG4-Biotin uses efficient, amine-specific NHS-ester chemistry and includes hydrophilic polyethylene glycol (PEG) spacer that stabilizes long-term solubility of labeled antibodies
• Eight biotin-labeling experiments per kit—Thermo Scientific No-Weigh Packaging ensures that the biotin reagent is fully active for eight separate experiments

This innovative antibody-labeling system uses nickel-chelated agarose to temporarily immobilize antibody molecules via their histidine-rich Fc regions. Once held in place on the resin, the antibody can be biotinylated at primary amines with the high-quality, solubility-stabilizing NHS-PEG4-Biotin reagent. Excess labeling reagent and byproducts are then washed away before recovering the labeled and purified antibody from the resin using a mild imidazole solution. No gel filtration or dialysis is needed.

EZ-Link Solid-Phase Biotinylation Kits use an innovative strategy for antibody biotinylation that simplifies the process in many respects, making successful biotinylation possible for the novice and expert. The strategy involves immobilizing the antibody to a metal-chelated affinity support and performing the biotinylation in the solid phase.

Antibodies bind readily to metal-chelated supports, such as nickel or cobalt, binding primarily through the histidine-rich Fc region of IgG-class antibodies. This feature allows the derivatization chemistry to be performed easily while the antibody is bound to the support. This strategy facilitates reagent delivery and removal of spent reagent. The resulting modified antibody is then selectively removed from the support by elution with a buffered imidazole solution.

The solid-phase approach to antibody biotinylation provides more control over the reaction. For example: Reagent and antibody ratios and reaction times can be adjusted to achieve the proper degree of biotinylation for the intended application.

The kits contain an aqueous soluble amino group-specific reagent (NHS-PEG4-Biotin) that targets accessible amino (-NH2) groups on the surface of the antibody. The format can be applied at the multi-milligram antibody level in the 1 mL column kit (Part No. 21440) or to low microgram antibody levels for biotinylations using the mini-spin column kit (Part No. 21450).

Related Products
EZ-Link™ NHS-PEG Solid-Phase Biotinylation Kit - 1 mL Column
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Labeling Scale0.5 to 1 mg
Product LineEZ-Link
Product TypeNHS-PEG Solid-Phase Biotinylation Kit
Quantity8 reactions
Labeling TargetIgG
Label or DyeBiotin
SpacerPegylated
Unit SizeEach
Contents & Storage
Sufficient For: 8 labeling reactions, each with 0.1 to 1 mg of antibody
• Ni-NTA Spin Columns, 0.2 mL, 8 columns
• PBS pack (makes 500 mL), 1 pack
• NHS-PEG4-Biotin, 8 x 2 mg
• Imidazole Stock Solution (4M), 5 mL
• Collection Tubes (2mL), 30 tubes

Store at 4°C.

Frequently asked questions (FAQs)

How do the EZ-Link Micro Biotinylation Kits differ from the standard biotinylation kits?

The standard biotinylation kits contain components for labeling 1-10 mg of protein, and the Micro Biotinylation Kits are structured for labeling 50-200 µg of protein in 200-700 mL. The Micro kits contain the biotinylation reagents in small, individual packages and do not include components to perform the HABA assay.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Can you provide the shelf-life for EZ-Link NHS-PEG Solid-Phase Biotinylation Kit?

EZ-Link NHS-PEG Solid-Phase Biotinylation Kit is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Citations & References (2)

Citations & References
Abstract
Identification of a novel HIV-1-neutralizing antibody from a CRF07_BC-infected Chinese donor.
Authors:Sun Y, Qiao Y, Zhu Y, Chong H, He Y
Journal:Oncotarget
PubMed ID:28968970
The identification of human monoclonal antibodies (mAbs) able to neutralize a broad spectrum of primary HIV-1 isolates is highly important for understanding the immune response of HIV-1 infection and developing vaccines and therapeutics. In this study, we isolated a novel human mAb termed Y498 from a phage display antibody library ... More
Regulatory T cells as a biomarker for response to adalimumab in rheumatoid arthritis.
Authors:Nguyen DX, Cotton A, Attipoe L, Ciurtin C, Doré CJ, Ehrenstein MR
Journal:J Allergy Clin Immunol
PubMed ID:29935955