CMRL Medium, no glutamine
Product Image
Gibco™

CMRL Medium, no glutamine

CMRL Medium was originally developed by Connaught Medical Research Laboratories for the growth of Earle's 'L'cells under serum-free conditions. Gibco™Read more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
21530027
also known as 21530-027
500 mL
21530076
also known as 21530-076
10 x 500 mL
Catalog number 21530027
also known as 21530-027
Price (INR)
-
Quantity:
500 mL
Customize this product

CMRL Medium was originally developed by Connaught Medical Research Laboratories for the growth of Earle's 'L'cells under serum-free conditions. Gibco™ CMRL Medium is also especially useful for cloning monkey kidney cells and for growth of many other mammalian cell lines when supplemented with horse or calf serum.

This CMRL Medium is modified as follows:

With: Phenol Red

Without: L-glutamine, HEPES

The complete formulation is available.

Gibco™ CMRL Medium is a modified version of Medium 199. CMRL medium is particularly rich in nucleosides and vitamins.

CMRL Medium contains no proteins or growth factors. Therefore, CMRL Medium requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). CMRL Medium uses a sodium bicarbonate buffer system (2.2 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.

Specifications
Cell TypeMammalian Cells
Concentration1 X
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Product LineGibco
Product TypeCMRL Medium
Quantity500 mL
Shelf Life12 Months
Shipping ConditionRoom Temperature
Culture TypeMammalian Cell Culture
FormLiquid
SterilitySterile-filtered
With AdditivesLow Glucose, Phenol Red
Without AdditivesNo Glutamine, No HEPES, No Sodium Pyruvate
Unit SizeEach
Contents & Storage
Storage conditions: 2-8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture

Citations & References (1)

Citations & References
Abstract
Tumor necrosis factor alpha increases the expression of glycosyltransferases and sulfotransferases responsible for the biosynthesis of sialylated and/or sulfated Lewis x epitopes in the human bronchial mucosa.
Authors: Delmotte Philippe; Degroote Sophie; Lafitte Jean-Jacques; Lamblin Geneviève; Perini Jean-Marc; Roussel Philippe;
Journal:J Biol Chem
PubMed ID:11679593
'There is increasing evidence that inflammation may affect glycosylation and sulfation of various glycoproteins. The present study reports the effect of tumor necrosis factor alpha (TNF-alpha), a proinflammatory cytokine, on the glycosyl- and sulfotransferases of the human bronchial mucosa responsible for the biosynthesis of Lewis x epitope and of its ... More