4-Acetophenetidide, 97%
4-Acetophenetidide, 97%
4-Acetophenetidide, 97%
Acros Organics

4-Acetophenetidide, 97%

CAS: 62-44-2 | C10H13NO2 | 179.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
500 g
Catalog number 216590025
also known as 21659
HSN 29251900
Price (INR)
30,039.00
Each
Quantity:
2.5 kg
Request bulk or custom format
Chemical Identifiers
CAS62-44-2
IUPAC NameN-(4-ethoxyphenyl)acetamide
Molecular FormulaC10H13NO2
InChI KeyCPJSUEIXXCENMM-UHFFFAOYSA-N
SMILESCCOC1=CC=C(NC(C)=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Infrared spectrumConforms
HPLC>=96.0 %
Melting point133°C to 138°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only