(R)-(-)-sec-Butanol, 99%
(R)-(-)-sec-Butanol, 99%
(R)-(-)-sec-Butanol, 99%
Acros Organics

(R)-(-)-sec-Butanol, 99%

CAS: 14898-79-4 | C4H10O | 74.123 g/mol
Have Questions?
Catalog NumberQuantityPackaging
217230010
also known as 21723
1 gGlass Bottle
Catalog number 217230010
also known as 21723
HSN 29051490
Price (INR)
9,225.00
Each
Quantity:
1 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (INR)
9,225.00
Each
Chemical Identifiers
CAS14898-79-4
IUPAC Name(2R)-butan-2-ol
Molecular FormulaC4H10O
InChI KeyBTANRVKWQNVYAZ-SCSAIBSYSA-N
SMILESCC[C@@H](C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
Specific optical rotation-12° to -14° (20°C, 589 nm) (neat)
Refractive index1.3960 to 1.3980 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only