Ruthenium(IV) oxide, 99.5+%, anhydrous
Ruthenium(IV) oxide, 99.5+%, anhydrous
Ruthenium(IV) oxide, 99.5+%, anhydrous
Ruthenium(IV) oxide, 99.5+%, anhydrous
Acros Organics

Ruthenium(IV) oxide, 99.5+%, anhydrous

CAS: 12036-10-1 | O2Ru | 133.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 218340050
HSN 28439019
Price (INR)
72,269.00
Each
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
72,269.00
Each
Add to cart
Chemical Identifiers
CAS12036-10-1
IUPAC Namedioxoruthenium
Molecular FormulaO2Ru
InChI KeyWOCIAKWEIIZHES-UHFFFAOYSA-N
SMILESO=[Ru]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Black
Purity>=99.50 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only