Deoxycholic acid, sodium salt, 99%, extra pure
Deoxycholic acid, sodium salt, 99%, extra pure
Deoxycholic acid, sodium salt, 99%, extra pure
Acros Organics

Deoxycholic acid, sodium salt, 99%, extra pure

Deoxycholic acid, sodium salt, CAS # 302-95-4, also designed as sodium deoxycholate, is a biological anionic bile acid detergent that is especially useful for disrupting and dissociating protein interactions. | CAS: 302-95-4 | C24H39NaO4 | 414.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 218590250
also known as 21859
HSN 29181990
Price (INR)
4,698.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS302-95-4
IUPAC Namesodium (4R)-4-[(1R,3aS,3bR,5aR,7R,9aS,9bS,11S,11aR)-7,11-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoate
Molecular FormulaC24H39NaO4
InChI KeyFHHPUSMSKHSNKW-SMOYURAASA-M
SMILES[Na+].[H][C@@]12CC[C@H]([C@H](C)CCC([O-])=O)[C@@]1(C)[C@@H](O)C[C@@]1([H])[C@@]2([H])CC[C@]2([H])C[C@H](O)CC[C@]12C
View more
SpecificationsSpecification SheetSpecification Sheet
pH7.5 to 10 (5 % in water)
Infrared spectrumConforms
Heavy metals (as Pb)=<20 ppm
TLC=<2 % Sodium cholate
Specific optical rotation+42° to +46° (20°C, 589 nm) (c=2, water)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • The deoxycholic acid sodium salt is a bile acid choleretic and anionic detergent that allows the solubilization of fats for intestinal absorption
  • The compound is an intermediate in the production of corticosteroids
  • In animal models it has been shown that this compound can promote tyrosine phosphorylation of β-catenin and its translocation to the nucleus, promoting the activation of β-catenin signaling


Applications

  • Deoxycholic acid is mainly useful for the extraction of membrane receptors, other plasma membrane proteins, nuclei isolation, solubilization and isolation of fats, and some components of biological membranes (in an active state)
  • It has been used for chemical applications, mainly for the elution or regeneration of specific types of affinity columns. Furthermore, this compound can be used to prevent non-specific binding in affinity chromatography.
  • It has been applied in the preparation of protein extracts for immunoblotting and immunoprecipitation of radiolabeled proteins
  • Deoxycholic acid, sodium salt is also recommended for stripping endotoxins
RUO – Research Use Only