Bacto™ Malt Extract
Gibco™
Bacto™ Malt Extract
Bacto Malt Extract is the water-soluble portion of malted barley. Due to the extraction process, this animal origin-free supplement providesRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
21861010 kg
218630500 g
Catalog number 218610
Price (INR)
93,756.00
Each
Quantity:
10 kg
Price (INR)
93,756.00
Each
Bacto Malt Extract is the water-soluble portion of malted barley. Due to the extraction process, this animal origin-free supplement provides a very high carbohydrate content to any medium. Bacto Malt Extract has been used successfully in microbial fermentation applications.
Specifications
Product LineBacto
Quantity10 kg
SourceAnimal origin-free, malted barley
FormPowder
Product TypeMedia Supplement
Unit SizeEach
Contents & Storage
Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.