Cesium, 99.95+%, (trace metal basis)
Cesium, 99.95+%, (trace metal basis)
Cesium, 99.95+%, (trace metal basis)
Acros Organics

Cesium, 99.95+%, (trace metal basis)

CAS: 7440-46-2 | Cs | 132.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number 219190050
also known as 21919
HSN 28051900
Price (INR)
136,395.00
Each
Quantity:
5 g
Request bulk or custom format
Price (INR)
136,395.00
Each
Chemical Identifiers
CAS7440-46-2
IUPAC Namecaesium
Molecular FormulaCs
InChI KeyTVFDJXOCXUVLDH-UHFFFAOYSA-N
SMILES[Cs]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Golden
Cesium (Cs)>=99.95 % (metals basis)
Total trace metal impurities=<500 ppm
Appearance (Form)Metal
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only