Cobalt(II) nitrate hexahydrate, 99+%, for analysis
Cobalt(II) nitrate hexahydrate, 99+%, for analysis
Cobalt(II) nitrate hexahydrate, 99+%, for analysis
Acros Organics

Cobalt(II) nitrate hexahydrate, 99+%, for analysis

CAS: 10026-22-9 | CoH12N2O12 | 291.03 g/mol
Have Questions?
Catalog NumberQuantityPackaging
219211000100 gPlastic bottle
Catalog number 219211000
HSN 28342920
Price (INR)
7,016.00
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
7,016.00
Each
Chemical Identifiers
CAS10026-22-9
IUPAC Nameλ²-cobalt(2+) hexahydrate dinitrate
Molecular FormulaCoH12N2O12
InChI KeyQGUAJWGNOXCYJF-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Adhering crystalline powder or crystals or chips
Heavy metals (as Cu)=<10 ppm
Iron (Fe)=<20 ppm
Zinc (Zn)=<50 ppm
Appearance (Color)Pink to red to brown
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
RUO – Research Use Only