Glycine, 99+%, for analysis
Glycine, 99+%, for analysis
Glycine, 99+%, for analysis
Acros Organics

Glycine, 99+%, for analysis

Glycine, CAS # 56-40-6, is a purified formulation of the amino acid, glycine, (aminoacetic acid) that is used in the synthesis of several compounds including creatinine, purines and porphyrins. | CAS: 56-40-6 | C2H5NO2 | 75.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 kg
100 g
250 g
1 kg
5 kg
Packaging:
View all
Fiber Drum
Catalog number 220910250
HSN 29224990
Price (INR)
175,619.00
Each
Quantity:
25 kg
Packaging:
Fiber Drum
Request bulk or custom format
Chemical Identifiers
CAS56-40-6
IUPAC Name2-aminoacetic acid
Molecular FormulaC2H5NO2
InChI KeyDHMQDGOQFOQNFH-UHFFFAOYSA-N
SMILESNCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO4>=99.0 % (on dried substance)
Residual solventsMeets the requirements
Arsenic (As)=<1 ppm
Appearance (Color)White
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

  • Glycine is an essential acid that can be used for synthesis of creatine, purines and porphyrins
  • Soluble in water; has a pH of ∼=5.5-6.5 in solution
  • Glycine is an inhibitory neurotransmitter that regulates the activity of NMDA receptors and affects neural activity

Applications

  • Glycine may also be used to maintain osmotic pressure, or as a component of other molecular biology buffers that are ion-dependent
  • Glycine is used in molecular biology as a buffer component for gel electrophoresis; the ability of glycine to adopt a negative charge at higher pH makes it ideal for separating proteins in the stacking layer
  • Intended for biochemistry applications
RUO – Research Use Only