Dionex™ Aquion™ RFIC System
Dionex™ Aquion™ RFIC System
Actual product may vary
Dionex™ Aquion™ RFIC System
Dionex™ Aquion™ RFIC System
Dionex™ Aquion™ RFIC System
Dionex™ Aquion™ RFIC System
Dionex™ Aquion™ RFIC System
Thermo Scientific™
Dionex™ Aquion™ RFIC System
Increase reliability, lower IC analysis cost, reduce error and save space with the automated eluent generation and compact design of the Dionex Aquion RFIC System.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionIncludes
22176-60128System w/ DegasChromeleon Software
22176-60017System w/o DegasSystem Only
22176-60018System w/ DegasSystem Only
22176-60125System w/o DegasChromeleon Software, PC
22176-60126System w/ DegasChromeleon Software, PC
22176-60127System w/o DegasChromeleon Software
Catalog number 22176-60128
Price (INR)
-
Request A Quote
Description:
System w/ Degas
Includes:
Chromeleon Software
Increase productivity and achieve confidence in your results with the Thermo Scientific™ Dionex™ Aquion™ RFIC system. This ion chromatography system is a simple, compact platform with eluent generation capabilities for budget-limited laboratories and features a dual-piston pump, electrolytic suppression, eluent generation, high sensitivity, and set-and-forget convenience. Eluent generation provides precise eluent concentration delivery that is programmed with the click of a mouse for unprecedented simplicity and error-free operation. This RFIC system is designed for ease-of-use, fast startup times, and reliable, stable performance for demanding applications in the environmental, food safety, and academic laboratories.
The Dionex Aquion RFIC System maintains low operating costs and incorporates eluent generation capabilities and electrolytic suppression to deliver results you can count on. Automated EG ensures better performance and reproducibility, and eliminates error-prone manual eluent preparation. This contamination-free, trouble-free RFIC features durable PEEK flow paths, which are compatible with the full range of IC eluents. The systems's reliable performance and reproducible results save you time and money.
Other system features and benefits include:
  • Increased productivity as a result of integrated electrolytic suppression and EG
  • Streamlined design with small footprint for minimal bench space
System standard features include:
  • Dual-piston pump
  • Eluent generation
  • Thermostatted digital conductivity cell
  • Electrolytic suppression
  • LED status panel
  • Standard bore and microbore column support
Available features include:
  • Column heater
  • In-line vacuum degas
  • Thermo Scientific™ Dionex™ Chromeleon™ Chromatography Data System (CDS) software
Specifications
DescriptionSystem w/ Degas
IncludesChromeleon Software
Detector TypeConductivity, Microprocessor-Controlled Digital Signal Processor
TypeIC System With Degas (Includes Chromeleon SW)
Unit SizeEach