Dionex™ ICS-6000 EG Eluent Generator
Thermo Scientific™

Dionex™ ICS-6000 EG Eluent Generator

Use the Thermo Scientific™ Dionex™ Eluent Generator in the ICS-6000 HPIC™ system to enable online electrolytic eluent generation using DI water and cartridges.

Have Questions?
Catalog NumberFlow Rate
22181-600190.001 to 10 mL/min.
Catalog number 22181-60019
Price (INR)
-
Flow Rate:
0.001 to 10 mL/min.

Minimize baselines, maximize consistency, and simplify your ion chromatography (IC) workflow with the Thermo Scientific™ Dionex™ ICS-6000 EG Eluent Generator. Add the EG module to a Thermo Scientific™ Dionex™ ICS-6000 HPIC™ system to create a Reagent-Free™ IC (RFIC™) system that produces single or dual eluents on-line electrolytically using only DI water and simple cartridges. Robust electronics generate high-purity eluents and exceptionally stable baselines. Just add water to generate the perfect eluent for almost any IC application—anion or cation, capillary or analytic, isocratic or gradient.

  • Generate high-purity eluents (up to 200mM for capillary systems) on demand
  • Run gradient separations as easily as isocratic separations
  • RFIC-EG systems combine “Just Add Water” eluent generation, eluent purification, and electrolytic suppression technologies
  • Provides superior performance, higher sensitivity, and excellent reproducibility while eliminating variability and potential contamination of systems using manually prepared eluents
Specifications
Depth (English)21.5 in.
Depth (Metric)56 cm
DescriptionDionex ICS-6000 EG Eluent Generator Dual (Does NOT include EG Degasser)
Electrical Requirements90 to 265 VAC 47/63 Hz
Frequency47/63 Hz
Gradient ProfilesAny Combination of an Unlimited Number of Linear, Convex, and Concave Positive and Negative Gradient Profiles
Height (English)16 in.
Height (Metric)41 cm
Max. Solvent ConcentrationCations: None; Anions: 25% Methanol
Min. Eluent Concentration0.01 mM
QuantityEach
Standard FeaturesBuilt-in leak sensor and operator alert; slide-out tray; Automated Procedure Wizards; System Smart Startup and Shutdown; System Wellness and Predictive Performance; Application Templates; Multivendor Automation Support of 3rd Party
Voltage90 to 265 VAC
Weight (English)40 lb.
Weight (Metric)25 kg
Width (English)8.75 in.
Width (Metric)23 cm
Flow Rate0.001 to 10 mL/min.
For Use With (Equipment)Dionex ICS-6000 HPIC System
Max Eluent Concentration100 mM (Analytical); 200 mM (Capillary)
Max. Pressure35 Mpa (5000 psi)
TypeEluent Generator
Unit SizeEach