(Bromomethyl)cyclopropane, 96%
(Bromomethyl)cyclopropane, 96%
(Bromomethyl)cyclopropane, 96%
Acros Organics

(Bromomethyl)cyclopropane, 96%

CAS: 7051-34-5 | C4H7Br | 135.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 221810010
also known as 22181
HSN 29035900
Price (INR)
5,634.00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
5,634.00
Each
Chemical Identifiers
CAS7051-34-5
IUPAC Name(bromomethyl)cyclopropane
Molecular FormulaC4H7Br
InChI KeyAEILLAXRDHDKDY-UHFFFAOYSA-N
SMILESBrCC1CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to slightly colored
Water=<0.1 % (K.F.)
Specific gravity(20°C) 1.385 to 1.415
Appearance (Form)Liquid
Refractive index1.4750 to 1.4790 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only