Diphenylamine, 99+%, extra pure, Redox-indicator
Diphenylamine, 99+%, extra pure, Redox-indicator
Diphenylamine, 99+%, extra pure, Redox-indicator
Acros Organics

Diphenylamine, 99+%, extra pure, Redox-indicator

CAS: 122-39-4 | C12H11N | 169.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 221921000
HSN 29214410
Price (INR)
8,119.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
8,119.00
Each
Chemical Identifiers
CAS122-39-4
IUPAC NameN-phenylaniline
Molecular FormulaC12H11N
InChI KeyDMBHHRLKUKUOEG-UHFFFAOYSA-N
SMILESN(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Sensitivity(to nitrate) Passes test
Appearance (Color)White to off-white
Sulfated ash=<0.05 %
Appearance (Form)Crystalline powder or crystals or flakes
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only