Pyridine hydrochloride, 98%, pure
Pyridine hydrochloride, 98%, pure
Pyridine hydrochloride, 98%, pure
Acros Organics

Pyridine hydrochloride, 98%, pure

CAS: 628-13-7 | C5H5N·ClH | 115.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 222285000
HSN 29333990
Price (INR)
20,381.00
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
20,381.00
Each
Chemical Identifiers
CAS628-13-7
IUPAC Namepyridin-1-ium chloride
Molecular FormulaC5H6ClN
InChI KeyAOJFQRQNPXYVLM-UHFFFAOYSA-N
SMILES[Cl-].C1=CC=[NH+]C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to tan
Titration Argentometric>=97.5 %
Solubilityalmost colorless
Infrared spectrumConforms
Solubility(5% in water) Clear colorless to
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only