Magnesium oxide, 97+%, for analysis
Magnesium oxide, 97+%, for analysis
Magnesium oxide, 97+%, for analysis
Acros Organics

Magnesium oxide, 97+%, for analysis

CAS: 1309-48-4 | MgO | 40.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
View all
Glass bottle
Catalog number 222532500
also known as 22253
HSN 25199090
Price (INR)
6,770.00
Each
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1309-48-4
IUPAC Nameoxomagnesium
Molecular FormulaMgO
InChI KeyCPLXHLVBOLITMK-UHFFFAOYSA-N
SMILESO=[Mg]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder
Titration Complexometric>=97.0 % (on dry substance)
Loss on ignition=<5.0 % (900°C)
Insoluble matter=<0.1 % (in HCl)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only