Bacto™ Casamino Acids
Gibco™
Bacto™ Casamino Acids
Bacto Casamino Acids is an animal origin (AO) acid hydrolysis of casein. This supplement contains low salt and iron contentRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
2230202 kg
22303010 kg
223050500 g
Catalog number 223020
Price (INR)
-
Quantity:
2 kg
Bacto Casamino Acids is an animal origin (AO) acid hydrolysis of casein. This supplement contains low salt and iron content making it an excellent supplement for media formulations where nitrogen requirements are minimal. This supplement is successfully used in fermentation applications for pneumococcal, diphtheria, pertussis, tetanus, and zoster vaccines. It maintains many of the nutritional benefits of hydrolyzed casein, but is further supplemented with growth factors, cystine, maltose, and an optimum amount of iron and inorganic salts.
Customers have used Bacto Casamino Acids to support the growth of Bordetella, Corynebacterium, Pseudomonas, Haemophilus, and Escherichia species. Additionally, together with Bacto Yeast Extract, technical, customers have used this supplement successfully in the production of animal and human health vaccines.
Specifications
FormatPail
Product LineGibco, Bacto
Quantity2 kg
Shelf Life5 Years
SourceAnimal Origin, Casein
SpeciesBovine
FormPowder
Product TypeDehydrated Culture Media
Regulatory StatusResearch Use or Further Manufacturing
Unit SizeEach
Contents & Storage
Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.