Potassium ferricyanide, 99+%, for analysis
Potassium ferricyanide, 99+%, for analysis
Potassium ferricyanide, 99+%, for analysis
Acros Organics

Potassium ferricyanide, 99+%, for analysis

CAS: 13746-66-2 | C6FeK3N6 | 329.25 g/mol
Have Questions?
Catalog NumberQuantityPackaging
223111000100 gGlass bottle
Catalog number 223111000
HSN 28372090
Price (INR)
5,807.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
5,807.00
Each
Chemical Identifiers
CAS13746-66-2
IUPAC Nametripotassium hexacyanoirontris(ylium)
Molecular FormulaC6FeK3N6
InChI KeyQMTKJUMXUDIUAQ-UHFFFAOYSA-N
SMILES[K+].[K+].[K+].N#C[Fe+3](C#N)(C#N)(C#N)(C#N)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Chloride (Cl)=<100 ppm
Appearance (Color)Orange to red
Appearance (Form)Crystals and/or chunks
Sulfate (SO4)=<100 ppm
Titration Iodometric>=99.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only