Sulfo-MBS (m-maleimidobenzoyl-N-hydroxysulfosuccinimide ester)
Thermo Scientific™

Sulfo-MBS (m-maleimidobenzoyl-N-hydroxysulfosuccinimide ester)

Thermo Scientific Pierce Sulfo-MBS is a water-soluble amine-to-sulfhydryl crosslinker that contains NHS-ester and maleimide reactive groups at opposite ends ofRead more
Have Questions?
Catalog NumberQuantity
2231250 mg
Catalog number 22312
Price (INR)
-
Quantity:
50 mg
Request bulk or custom format
Thermo Scientific Pierce Sulfo-MBS is a water-soluble amine-to-sulfhydryl crosslinker that contains NHS-ester and maleimide reactive groups at opposite ends of a short aromatic spacer arm (7.3 angstroms).

Features of Sulfo-MBS:

Reactive groups: sulfo-NHS ester and maleimide
Reactive towards: amino and sulfhydryl groups
• Non-cleavable
• Water-soluble (compare to MBS)
• Membrane impermeable, allowing cell surface labeling

Product References:
Crosslinker Application Guide -- search for recent literature references for this product

Related Products
MBS (m-maleimidobenzoyl-N-hydroxysuccinimide ester)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell PermeabilityNo
DescriptionSulfo-MBS
FormPowder
Labeling MethodChemical Labeling
Molecular Weight (g/mol)416.29
PEGylatedNo
Product LinePierce
Quantity50 mg
Reactive MoietyMaleimide, Sulfo-NHS Ester
Shipping ConditionAmbient
SolubilityWater
Spacer Arm Length7.3 Å
Water SolubleYes
Chemical ReactivityAmine-Sulfhydryl
CleavableNo
Crosslinker TypeHeterobifunctional
FormatStandard
Product TypeCrosslinker
SpacerShort (<10 Å)
Unit SizeEach
Contents & Storage
Upon receipt store desiccated at -20°C.

Frequently asked questions (FAQs)

Can you provide the shelf-life for Sulfo-MBS (m-maleimidobenzoyl-N-hydroxysulfosuccinimide ester)?

Sulfo-MBS (m-maleimidobenzoyl-N-hydroxysulfosuccinimide ester) is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Citations & References (3)

Citations & References
Abstract
Solid phase DNA amplification: characterisation of primer attachment and amplification mechanisms.
Authors:Adessi C, Matton G, Ayala G, Turcatti G, Mermod JJ, Mayer P, Kawashima E
Journal:Nucleic Acids Res
PubMed ID:11024189
Different chemical methods used to attach oligonucleotides by their 5'-end on a glass surface were tested in the framework of solid phase PCR where surface-bound instead of freely-diffusing primers are used to amplify DNA. Each method was first evaluated for its capacity to provide a high surface coverage of oligonucleotides ... More
Interaction with the 5D3 monoclonal antibody is regulated by intramolecular rearrangements but not by covalent dimer formation of the human ABCG2 multidrug transporter.
Authors:Ozvegy-Laczka C, Laczkó R, Hegedus C, Litman T, Várady G, Goda K, Hegedus T, Dokholyan NV, Sorrentino BP, Váradi A, Sarkadi B
Journal:J Biol Chem
PubMed ID:18644784
Human ABCG2 is a plasma membrane glycoprotein working as a homodimer or homo-oligomer. The protein plays an important role in the protection/detoxification of various tissues and may also be responsible for the multidrug-resistant phenotype of cancer cells. In our previous study we found that the 5D3 monoclonal antibody shows a ... More
Rat liver carnitine palmitoyltransferase 1 forms an oligomeric complex within the outer mitochondrial membrane.
Authors:Faye A, Esnous C, Price NT, Onfray MA, Girard J, Prip-Buus C
Journal:J Biol Chem
PubMed ID:17650509
Carnitine palmitoyltransferase (CPT) 1A catalyzes the rate-limiting step in the transport of long chain acyl-CoAs from cytoplasm to the mitochondrial matrix by converting them to acylcarnitines. Located within the outer mitochondrial membrane, CPT1A activity is inhibited by malonyl-CoA, its allosteric inhibitor. In this study, we investigate for the first time ... More