Bacto™ Casamino Acids, technical
Gibco™

Bacto™ Casamino Acids, technical

Bacto Casamino Acids, technical, is an animal origin (AO) acid hydrolysis of casein. In contrast to the Bacto Casamino Acids,Read more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
223120500 g
22311010 kg
Catalog number 223120
Price (INR)
-
Quantity:
500 g
Bacto Casamino Acids, technical, is an animal origin (AO) acid hydrolysis of casein. In contrast to the Bacto Casamino Acids, this supplement has been processed less and, as a result, has higher sodium chloride and iron content. Customers have successfully used Bacto Casamino Acids, technical, in the production of a number of animal health vaccines, and it has been shown to support the growth of Bordetella, Haemophilus, and Vibrio species.

Specifications
Certifications/ComplianceAOAC, BAM, COMPF, SMWW, USDA, USP, ISO/DIS, 11014
For Use With (Application)Animal health vaccines, support the growth of Bordetella, Haemophilus, and Vibrio species
FormatBottle
Product LineBacto
Purity or Quality GradeTechnical Grade
Quantity500 g
Shelf Life1825 Days
SourceAnimal origin, casein
FormPowder
Product TypeCasamino Acids
Unit SizeEach
Contents & Storage
Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.