Sodium perborate tetrahydrate, 97%, pure
Sodium perborate tetrahydrate, 97%, pure
Sodium perborate tetrahydrate, 97%, pure
Sodium perborate tetrahydrate, 97%, pure
Acros Organics

Sodium perborate tetrahydrate, 97%, pure

CAS: 10486-00-7 | BNaO3·4H2O | 153.86 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
250 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number 223200250
also known as 22320
HSN 28403000
Price (INR)
5,231.00
Each
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
5,231.00
Each
Chemical Identifiers
CAS10486-00-7
IUPAC Namedisodium tetrahydroxy-1,2,4,5,3,6-tetraoxadiborinane-3,6-diuide hexahydrate
Molecular FormulaB2H16Na2O14
InChI KeyMCYPYXIRGFEFGL-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.[Na+].[Na+].O[B-]1(O)OO[B-](O)(O)OO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline or granular powder or crystals
Titration with KMnO496.0 to 104.0 %
pH10.1 to 10.6 (1 % solution)

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only