Medium 199, Hanks' Balanced Salts
Product Image
Gibco™

Medium 199, Hanks' Balanced Salts

Medium 199 was originally developed for nutritional studies of chick embryo fibroblasts. It has broad species applicability, particularly for cultivationRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
22350078
also known as 22350-078
10 x 500 mL
22350029
also known as 22350-029
500 mL
Catalog number 22350078
also known as 22350-078
Price (INR)
-
Quantity:
10 x 500 mL
Customize this product

Medium 199 was originally developed for nutritional studies of chick embryo fibroblasts. It has broad species applicability, particularly for cultivation of non-transformed cells. Medium 199 is widely used in virology, vaccine production, and in vitro cultivation of primary explants of mouse pancreatic epithelium, and rat lens tissues. We offer a variety of Gibco™ Medium 199 modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This Medium 199 is modified as follows:

With: Hanks' salts, L-glutamine, Bacto-peptone, HEPES, Phenol Red

The complete formulation is available.

Compared to other basal media, Medium 199 contains unique components, including adenine, adenosine, hypoxanthine, thymine, and additional vitamins. This Medium 199 formulation contains Hanks' salts for use without CO2.

Medium 199 contains no proteins, lipids, or growth factors. Therefore, MEM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). This Medium 199 formulation does not require a CO2 environment to maintain physiological pH.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Cell TypeMammalian Cells
Concentration1 X
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Product LineGibco
Product TypeMedium 199
Quantity10 x 500 mL
Shelf Life12 Months
Culture TypeMammalian Cell Culture
FormLiquid
SterilitySterile-filtered
With AdditivesGlutamine, HEPES, Phenol Red, Hanks’ Balanced Salt solution (HBSS)
Unit SizeEach
Contents & Storage
Storage conditions: 2-8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture