4-Methylsalicylic acid, 98%
4-Methylsalicylic acid, 98%
4-Methylsalicylic acid, 98%
Acros Organics

4-Methylsalicylic acid, 98%

CAS: 50-85-1 | C8H8O3 | 152.149 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 223540250
also known as 22354
HSN 29182910
Price (INR)
7,883.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
7,883.00
Each
Chemical Identifiers
CAS50-85-1
IUPAC Name2-hydroxy-4-methylbenzoic acid
Molecular FormulaC8H8O3
InChI KeyNJESAXZANHETJV-UHFFFAOYSA-N
SMILESCC1=CC=C(C(O)=O)C(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point171°C to 179°C
Titration with NaOH>=97.5 %
Appearance (Color)White to grey-beige
Infrared spectrumConforms
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only