Potassium, 98%, chunks, in mineral oil
Potassium, 98%, chunks, in mineral oil
Potassium, 98%, chunks, in mineral oil
Acros Organics

Potassium, 98%, chunks, in mineral oil

CAS: 7440-09-7 | K | 39.10 g/mol
Have Questions?
Catalog NumberQuantityPackaging
223865000500 gPlastic Drum
Catalog number 223865000
HSN 28051900
Price (INR)
97,526.00
Each
Quantity:
500 g
Packaging:
Plastic Drum
Request bulk or custom format
Price (INR)
97,526.00
Each
Chemical Identifiers
CAS7440-09-7
IUPAC Namepotassium
Molecular FormulaK
InChI KeyZLMJMSJWJFRBEC-UHFFFAOYSA-N
SMILES[K]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Grey
Total trace metal impurities=<2 %
Appearance (Form)Chunks
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only