Product Image
Applied Biosystems™

Tube, 1/4 in. OD Polypropylene (for the Procise™ Protein Sequencer)

This is a Procise Protein Sequencer Spare Part.
Have Questions?
Catalog NumberQuantity
2250161 Unit
Catalog number 225016
Price (INR)
-
Quantity:
1 Unit

This is a Procise Protein Sequencer Spare Part.

For Research Use Only. Not for use in diagnostics procedures.

Specifications
For Use With (Equipment)Procise Protein Sequencer
Quantity1 Unit
Shipping ConditionRoom Temperature
Product TypeProtein Sequencer Tube
Unit SizeEach
Contents & Storage
Store at room temperature.