gamma-Benzyl L-glutamate, 99%
gamma-Benzyl L-glutamate, 99%
gamma-Benzyl L-glutamate, 99%
Acros Organics

gamma-Benzyl L-glutamate, 99%

CAS: 1676-73-9 | C12H15NO4 | 237.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Packaging:
Glass bottle
Catalog number 225670050
HSN 29224990
Price (INR)
9,090.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1676-73-9
IUPAC Name2-amino-5-(benzyloxy)-5-oxopentanoic acid
Molecular FormulaC12H15NO4
InChI KeyBGGHCRNCRWQABU-UHFFFAOYNA-N
SMILESNC(CCC(=O)OCC1=CC=CC=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=98.5 %
Infrared spectrumConforms
Specific optical rotation+26° to +29° (20°C, 589 nm) (c=2, 1N HCl)
Appearance (Form)Powder
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only