6-Chloropurine riboside, 99+%
6-Chloropurine riboside, 99+%
6-Chloropurine riboside, 99+%
Acros Organics

6-Chloropurine riboside, 99+%

CAS: 5399-87-1 | C10H11ClN4O4 | 286.67 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 226450050
also known as 22645
HSN 32049000
Price (INR)
39,957.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
39,957.00
Each
Chemical Identifiers
CAS5399-87-1
IUPAC Name2-(6-chloro-9H-purin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
Molecular FormulaC10H11ClN4O4
InChI KeyXHRJGHCQQPETRH-UHFFFAOYNA-N
SMILESOCC1OC(C(O)C1O)N1C=NC2=C(Cl)N=CN=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Infrared spectrumConforms
HPLC>=99.0 %
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only