6-Mercaptopurine monohydrate, 98%
6-Mercaptopurine monohydrate, 98%
6-Mercaptopurine monohydrate, 98%
6-Mercaptopurine monohydrate, 98%
Acros Organics

6-Mercaptopurine monohydrate, 98%

CAS: 6112-76-1 | C5H6N4OS | 170.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 226520050
HSN 29335990
Price (INR)
10,463.00
Each
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
10,463.00
Each
Add to cart
Chemical Identifiers
CAS6112-76-1
IUPAC Name6,7-dihydro-3H-purine-6-thione hydrate
Molecular FormulaC5H6N4OS
InChI KeyWFFQYWAAEWLHJC-UHFFFAOYSA-N
SMILESO.S=C1N=CNC2=C1NC=N2
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only