beta-Cyclodextrin hydrate, 99%
beta-Cyclodextrin hydrate, 99%
beta-Cyclodextrin hydrate, 99%
Acros Organics

beta-Cyclodextrin hydrate, 99%

CAS: 68168-23-0 | C42H70O35 | 1134.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 227280050
also known as 22728
HSN 29400000
Price (INR)
2,722.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
2,722.00
Each
Chemical Identifiers
CAS68168-23-0
IUPAC Name5,10,15,20,25,30,35-heptakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34-tetradecaoxaoctacyclo[31.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹]nonatetracontane-36,37,38,39,40,41,42,43,44,45,46,47,48,49-tetradecol
Molecular FormulaC42H70O35
InChI KeyWHGYBXFWUBPSRW-UHFFFAOYSA-N
SMILESOCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC8C(CO)OC(OC1C(O)C2O)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O
View more
SpecificationsSpecification SheetSpecification Sheet
Specific optical rotation+155° to +167° (25°C, 589 nm) (c=1, water)
Specific optical rotationOn dry substance
pH5.0 to 8.0 (1 % in water)
Residual solvents=<5 ppm Toluene
Reducing substances=<0.2 % (as Glucose)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagent
RUO – Research Use Only