Nalpha-Acetyl-L-lysine, 99+%
Nalpha-Acetyl-L-lysine, 99+%
Nalpha-Acetyl-L-lysine, 99+%
Acros Organics

Nalpha-Acetyl-L-lysine, 99+%

CAS: 1946-82-3 | C8H16N2O3 | 188.23 g/mol
Have Questions?
Catalog NumberQuantityPackaging
227630010
also known as 22763
1 gGlass bottle
Catalog number 227630010
also known as 22763
HSN 29241900
Price (INR)
17,612.00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
17,612.00
Each
Chemical Identifiers
CAS1946-82-3
IUPAC Name(2S)-6-amino-2-acetamidohexanoic acid
Molecular FormulaC8H16N2O3
InChI KeyVEYYWZRYIYDQJM-JLDDOWRYNA-N
SMILESCC(=O)N[C@@H](CCCCN)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with HClO499.0 to 101.0 %
Water=<1.0 % (K.F.)
Specific optical rotation+5.0° to +6.0° (20°C, 589 nm) (c=1, H2O)
Appearance (Color)White to almost white
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only