Chenodeoxycholic acid, 98%
Chenodeoxycholic acid, 98%
Chenodeoxycholic acid, 98%
Acros Organics

Chenodeoxycholic acid, 98%

Chenodeoxycholic acid, 98%, ACROS Organics, Quantity: 5g, Packaging: Glass bottle, White, Melting Point: 162 deg.C to 168 deg.C, Molecular Weight: 391.572, Percent Purity: >-97.5%, Assay Percent Range: 97+%, CAS: 474-25-9, ChEBI: CHEBI:36234, EINECS Number: 207-481-8, Formula Weight: 392.57
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Packaging:
Glass bottle
Catalog number 228770250
also known as 22877
HSN 29181990
Price (INR)
40,857.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
40,857.00
Each
Chemical Identifiers
CAS474-25-9
IUPAC Name(4R)-4-[(1R,3aS,3bR,4R,5aS,7R,9aS,9bS,11aR)-4,7-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoic acid
Molecular FormulaC24H40O4
InChI KeyRUDATBOHQWOJDD-BSWAIDMHSA-N
SMILES[H][C@@]12CC[C@H]([C@H](C)CCC(O)=O)[C@@]1(C)CC[C@@]1([H])[C@@]2([H])[C@H](O)C[C@]2([H])C[C@H](O)CC[C@]12C
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to off-white
Appearance (Form)Powder
Titration with NaOH>=97.0 % (on dried basis)
Loss on drying=<2 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only