Auto-refill for VersaCool™ Refrigerated Circulating Bath
Auto-refill for VersaCool™ Refrigerated Circulating Bath
Auto-refill for VersaCool™ Refrigerated Circulating Bath
Thermo Scientific™
Auto-refill for VersaCool™ Refrigerated Circulating Bath
Automatically maintain and ensure the optimal level of fluid in your VersaCool with the Autorefill feature.
Have Questions?
Catalog NumberFor Use With (Equipment)
230000005Thermo Scientific VersaCool Refrigerated Circulating Bath
Catalog number 230000005
Price (INR)
-
For Use With (Equipment):
Thermo Scientific VersaCool Refrigerated Circulating Bath
Automatically maintain and ensure the optimal level of fluid in your VersaCool with the Autorefill feature. Provides peace of mind with user-defined start and stop levels, as well as a time-out feature. The needle valve allows control of the fill rate from as fast a refill as possible- to a slow refill that minimizes any effect on temperature stability or any rate in between
Specifications
DescriptionAuto-refill for VersaCoolandtrade Refrigerated Circulating Bath
For Use With (Equipment)Thermo Scientific VersaCool Refrigerated Circulating Bath
TypeAuto-refill Device
Unit SizeEach