Nunc™ Strips and Frames
Actual product may vary
Thermo Scientific™
Nunc™ Strips and Frames
Maximize heat transmission and sample recovery using Nunc Strips and Frames.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct Type
232042Sequencing Module
232298Frame for Module
Catalog number 232042
Price (INR)
-
Product Type:
Sequencing Module
Maximize heat transmission and sample recovery using Thermo Scientific™ Nunc Strips and Frames. Thermo Scientific Nunc Life Science products offer reliability and reproducible results, and are backed by industry-leading quality standards and certifications.
  • Tape and well design reduce evaporation
  • Centrifugation eliminated when mixing reagents
  • Easy well identification
  • Minimum binding of hydrophilic molecules
  • Standard 96 well format
  • Modules are temperature resistant from -70° to +121°C
  • Space saving at the bench and in the freezer
  • Plastic waste is reduced
Specifications
FormatStrip Plate
Product TypeSequencing Module
Array8 x 12
MaterialPolypropylene
No. of Wells96
Unit SizeEach