Pierce™ Bradford Plus Protein Assay Reagent
Thermo Scientific™

Pierce™ Bradford Plus Protein Assay Reagent

The Pierce Bradford Plus Protein Assay is a ready-to-use, reducing agent-compatible, improved Bradford assay reagent to quickly measure total proteinRead more
Have Questions?
Catalog NumberQuantity
23238300 mL
Catalog number 23238
Price (INR)
24,889.00
Online offer
Ends: 31-Dec-2026
35,555.00
Save 10,666.00 (30%)
Each
Quantity:
300 mL
Request bulk or custom format
Price (INR)
24,889.00
Online offer
Ends: 31-Dec-2026
35,555.00
Save 10,666.00 (30%)
Each
The Pierce Bradford Plus Protein Assay is a ready-to-use, reducing agent-compatible, improved Bradford assay reagent to quickly measure total protein concentration compared to a protein standard. The Pierce Bradford Plus Assay Reagent provides increased linearity and half the protein-to-protein variability of other commercial Bradford assay formulations.

Compare all available Bradford assays ›

Features of the Bradford Plus Protein Assay include:
• Colorimetric—measure with a standard spectrophotometer or plate reader at 595 nm
• Easy to use—single reagent; no working reagent preparation required
• Fast—almost immediate color development; add, mix, and read results
• Assay range—detects protein concentration in the range 1 to 1500 μg/mL
• Better—improved linearity and response uniformity compared to traditional Bradford formulations
• Flexible—microplate and cuvette protocols provided and adaptable to several target working ranges

The Pierce Bradford Plus Assay Reagent is a single, ready-to-use solution for measuring protein concentration. Simply add the reagent to equal volumes of samples and standards, mix, and then measure the absorbance at 595 nm. The assay can be performed in either test tube or microplate format. The protein assay is compatible with most salts, solvents, buffers, thiols, reducing substances, and metal chelating agents encountered in protein samples.

How the Bradford Plus Assay detects protein
Use of Coomassie G-250 dye in a colorimetric reagent for the detection and quantitation of total protein was first described by Dr. Marion Bradford in 1976. In the acidic environment of the reagent, protein binds to the Coomassie dye. This results in a spectral shift from the reddish/brown form of the dye (absorbance maximum at 465 nm) to the blue form of the dye (absorbance maximum at 610 nm). The difference between the two forms of the dye is greatest at 595 nm, so that is the optimal wavelength to measure the blue color from the Coomassie dye-protein complex. If desired, the blue color can be measured at any wavelength between 575 nm and 615 nm. At the two extremes (575 nm and 615 nm) there is a loss of about 10% in the measured amount of color (absorbance) compared to that obtained at 595 nm.

Development of color in Coomassie dye-based (Bradford) protein assays has been associated with the presence of certain basic amino acids (primarily arginine, lysine, and histidine) in the protein. Van der Waals forces and hydrophobic interactions also participate in the binding of the dye by protein. The number of Coomassie dye ligands bound to each protein molecule is approximately proportional to the number of positive charges found on the protein. Free amino acids, peptides, and low molecular weight proteins do not produce color with Coomassie dye reagents. In general, the mass of a peptide or protein must be at least 3,000 daltons to be assayed with this reagent.

Related products
Pierce Bradford Plus Assay Kit
Pierce Detergent Compatible Bradford Assay Kit
Pierce Dilution-Free BSA Protein Standards, Multichannel Pipette Compatible, 0.125–2 mg/mL

For Research Use Only. Not for use in diagnostic procedures.
Specifications
AssayBradford Assay
DescriptionPierce Bradford Plus Protein Assay Reagent
For Use With (Application)Solution-based Detection, Absorbance
For Use With (Equipment)Spectrophotometer, Microplate Reader
Product LinePierce
Product TypeProtein Quantitation Assay
Quantity300 mL
SpecificityNot Target-Specific
Detection MethodColorimetric
Unit SizeEach
Contents & Storage
Store at 4°C.

Frequently asked questions (FAQs)

What is Pierce Bradford Plus Protein Assay Reagent?

Pierce Bradford Plus Protein Assay Reagent is a quick and ready-to-use modification of the well-known Bradford, Coomassie dye-binding, colorimetric method for total protein quantitation. This Bradford reagent modification greatly reduces the tendency of Coomassie dye-containing reagents to give a characteristically nonlinear response curve. Pierce Bradford Plus Protein Assay Reagent results in a substantially more linear response curve in a defined range of protein concentration. Thermo Scientific's special formulation results in significantly less protein-to-protein variation than that which is observed with other Bradford-type Coomassie dye-based formulations. When Coomassie dye binds protein in an acidic medium, an immediate absorbance shift occurs from 465 nm to 595 nm, with a simultaneous color change of the reagent from green/brown (or red/brown) to blue.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

How can I remove interfering substances from my sample before using the Pierce Bradford Plus Protein Assay Reagent?

You can remove interfering substances easily with the use of one of several Thermo Scientific products designed to purify your sample. These products include Slide-A-Lyzer Dialysis Cassettes, D-Salt Desalting Columns and Extracti-Gel D Detergent Removing Gel. Furthermore, TCA and cold acetone can be utilized to remove other interfering substances. (See the TechTips section of this website for more information.)

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Can detergents be used to clean my glassware interfere with the Pierce Bradford Plus Protein Assay Reagent?

Care must be exercised when cleaning glassware that will be used again for protein assays. Thorough cleaning often requires the use of a detergent but it must be completely removed in the final rinse. Also, the Coomassie dye will stain glass or quartz cuvettes. The cuvettes can be cleaned easily with our detergents followed by a thorough final rinsing with deionized water. If you prefer, disposable polystyrene cuvettes may be used to eliminate the cuvette-cleaning chore.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Can I read at wavelengths other than 595 nm with the Pierce Bradford Plus Protein Assay Reagent?

If a photometer or plate reader is not available with a 595 nm filter, the blue color may be measured at any wavelength between 570 nm and 610 nm. The maximum sensitivity of the assay occurs when the absorbance of the Coomassie dye protein complex is measured at 595 nm. Taking the absorbance measurements at any wavelength other than 595 nm will result in a lower slope for the standard curve and may increase the minimum detection level for the protocol.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.