Product Image
Thermo Scientific Chemicals

Chloral hydrate, 98.5%

CAS: 302-17-0 | C2H3Cl3O2 | 165.39 g/mol
Have Questions?
Catalog NumberQuantity
232475000500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 232475000
Price (INR)
-
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS302-17-0
IUPAC Name2,2,2-trichloroethane-1,1-diol
Molecular FormulaC2H3Cl3O2
InChI KeyRNFNDJAIBTYOQL-UHFFFAOYSA-N
SMILESOC(O)C(Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white
Appearance (Form)Powder or crystals
Infrared spectrumConforms
Titration excess NaOH98.0 to 102.0 %
Chloride (Cl)=<200 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only