Hexamethylenetetramine, 98.5%, contains an anticaking agent
Hexamethylenetetramine, 98.5%, contains an anticaking agent
Hexamethylenetetramine, 98.5%, contains an anticaking agent
Acros Organics

Hexamethylenetetramine, 98.5%, contains an anticaking agent

CAS: 100-97-0 | C6H12N4 | 140.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
2.5 kg
Packaging:
Plastic bottle
Catalog number 232890010
HSN 29336990
Price (INR)
6,226.00
Each
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
6,226.00
Each
Chemical Identifiers
CAS100-97-0
IUPAC Name1,3,5,7-tetraazatricyclo[3.3.1.1³,⁷]decane
Molecular FormulaC6H12N4
InChI KeyVKYKSIONXSXAKP-UHFFFAOYSA-N
SMILESC1N2CN3CN1CN(C2)C3
View more
SpecificationsSpecification SheetSpecification Sheet
Titration excess HCl>=98.0 %
Appearance (Color)White
Appearance (Form)Crystalline powder
Water=<0.5 % (K.F.)
Ash0.5 % to 3.0 % (anticaking agent)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only