Iron(II) sulfide, fused sticks
Iron(II) sulfide, fused sticks
Iron(II) sulfide, fused sticks
Acros Organics

Iron(II) sulfide, fused sticks

CAS: 1317-37-9 | FeS | 87.91 g/mol
Have Questions?
Catalog number 233300010
HSN 28309010
Price (INR)
9,918.00
Each
Request bulk or custom format
Price (INR)
9,918.00
Each
Chemical Identifiers
CAS1317-37-9
IUPAC Nameλ²-iron(2+) sulfanediide
Molecular FormulaFeS
InChI KeyGNVXPFBEZCSHQZ-UHFFFAOYSA-N
SMILES[S--].[Fe++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Bluish-black to grey-black
Appearance (Form)Sticks
WidthDiameter about 1-1.5 cm
Assay>=25 % (Sulfide (S))
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only