PFHM-II Protein-Free Hybridoma Medium (powder)
PFHM-II Protein-Free Hybridoma Medium (powder)
PFHM-II Protein-Free Hybridoma Medium (powder)
Gibco™

PFHM-II Protein-Free Hybridoma Medium (powder)

Gibco® Protein Free Hybridoma Medium II (PFHM II) is a serum-free, protein-free medium specifically developed for the ability to supportRead more
Have Questions?
Catalog NumberQuantity
236000421 x 10 L
Catalog number 23600042
Price (INR)
35,589.00
Online offer
Ends: 31-Mar-2026
52,724.00
Save 17,135.00 (32%)
Each
Quantity:
1 x 10 L
Price (INR)
35,589.00
Online offer
Ends: 31-Mar-2026
52,724.00
Save 17,135.00 (32%)
Each
Gibco® Protein Free Hybridoma Medium II (PFHM II) is a serum-free, protein-free medium specifically developed for the ability to support the growth of hybridomas for antibody production. Gibco® PFHM II features:
• Ability to support suspension and stationary hybridomas and myelomas
• Complete, ready-to-use formulation
• Serum-free, protein-free formulation
• Provided in a convenient easy-to-reconstitute powder format

Ability to support suspension and stationary hybridomas and myelomas
Suitable for the culture of recombinant myeloma lines as well as traditional hybridomas, Gibco® PFHM II provides high cell densities and IgG production. Cholesterol-dependent cultures such as NS0 require the addition of 250x Cholesterol Lipid Concentrate.

Complete, ready-to-use formulation
Once reconstituted, Gibco® PFHM II is a complete medium that is ready-to-use. No supplementation is required.

Serum-free, very low-protein formulation
Gibco® PFHM II is a serum-free, protein-free formulation, allowing for easier purification of your protein of interest.

Product Use
For Research Use Only. Not for use in diagnostic procedures.

cGMP Manufacturing and Quality System
Gibco® PFHM II is manufactured at a cGMP compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeHybridomas
Product LineGibco
Product TypeHybridoma Medium, powder
Quantity1 x 10 L
Shelf Life24 Months
Shipping ConditionRoom Temperature
ClassificationProtein-free
Culture TypeSuspension Cell Culture
FormPowder
Serum LevelSerum-free
With AdditivesGlutamine, Phenol Red
Unit SizeEach
Contents & Storage
Storage conditions: 2-8°C
Shipping conditions: Ambient
Shelf life: 24 months from date of manufacture