Nunc™ Seals, Tapes, and Foils
Actual product may vary
Thermo Scientific™

Nunc™ Seals, Tapes, and Foils

Minimize evaporation and protect samples from contamination and spilling with these sealing tapes, ideal for all assay applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMaterialFor Use With (Application)Certified RNase/DNase-free
236710Polyolefin AcrylateLow autofluorescenceYes
236366PKPolyesterCell culture, sterile applicationsNo
5701TS1PolyesterShort-term storage, ELISA, minimizes evaporation and cross contaminationNo
Catalog number 236710
Price (INR)
-
Material:
Polyolefin Acrylate
For Use With (Application):
Low autofluorescence
Certified RNase/DNase-free:
Yes

Minimize evaporation and protect samples from contamination and spilling with Thermo Scientific™ Nunc™ Sealing Tapes. These adhesive seals are ideal for all assay applications. Nunc Life Science products offer reliability and reproducible results, and are backed by industry-leading quality standards and certifications.

  • Adhesive seals effectively seal all microplate formats
  • Pressure sensitive adhesive is not tacky to touch, but forms a strong bond to the plate upon application of pressure
  • Thin, lightweight tapes seal onto the plate with a convenient, handheld applicator (available separately, Cat. No. 250050)
Specifications
For Use With (Application)Low autofluorescence
No. per Case400
No. per Pack400
Product TypeFilm, Seal, or Applicator
Solvent Or Alcohol ResistantDMSO Resistant
Temperature Range (Metric)-70°C to +100°C
AdhesivePressure Sensitive
Certified RNase/DNase-freeYes
ColorClear
MaterialPolyolefin Acrylate
PierceableNo
Product LineNunc
Unit SizeEach

Frequently asked questions (FAQs)

Can I boil Nunc Sealing Tape?

Our Nunc Stealing Tape is thermostable from -20 degrees C to 100 degrees C.