Search
| Catalog Number | Capacity (Metric) | Closure Size |
|---|---|---|
| 2402-0250PK also known as 2402-0250 | 250 mL | 24 mm |
| 2402-0500PK also known as 2402-0500 | 500 mL | 38 mm |
| 2402-1000PK also known as 2402-1000 | 1000 mL | 43 mm |
Dispense contents without tipping or shaking with Thermo Scientific™ Nalgene™ Unitary™ LDPE Wash Bottles. Wide mouth allows faster, easier filling. Bottle and tubulation are molded in one piece for uniform service and precision flow without sputtering. The tip can be cut back to increase flow. Made from high-quality, laboratory-grade, plastic materials for dependably low leachables and extractables. Naturally flexible LDPE material is chemically compatible with a wide range of laboratory reagents and provides an ergonomic soft squeeze to help reduce hand fatigue.
Features:
Includes:
Polypropylene screw closure.
Recycling information:
Nalgene reusable LDPE wash bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Nalgene LDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene LDPE bottles.