Nalgene™ Unitary™ LDPE Wash Bottles
Nalgene™ Unitary™ LDPE Wash Bottles
Nalgene™ Unitary™ LDPE Wash Bottles
Nalgene™ Unitary™ LDPE Wash Bottles
Nalgene™ Unitary™ LDPE Wash Bottles
Thermo Scientific™

Nalgene™ Unitary™ LDPE Wash Bottles

Dispense contents without tipping or shaking by using these LDPE wash bottles, designed with a wide mouth for faster, easier filling.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure Size
2402-0500PK
also known as 2402-0500
500 mL38 mm
2402-0250PK
also known as 2402-0250
250 mL24 mm
2402-1000PK
also known as 2402-1000
1000 mL43 mm
Catalog number 2402-0500PK
also known as 2402-0500
Price (INR)
11,879.00
Pack of 4
Add to cart
Capacity (Metric):
500 mL
Closure Size:
38 mm
Price (INR)
11,879.00
Pack of 4
Add to cart

Dispense contents without tipping or shaking with Thermo Scientific™ Nalgene™ Unitary™ LDPE Wash Bottles. Wide mouth allows faster, easier filling. Bottle and tubulation are molded in one piece for uniform service and precision flow without sputtering. The tip can be cut back to increase flow. Made from high-quality, laboratory-grade, plastic materials for dependably low leachables and extractables. Naturally flexible LDPE material is chemically compatible with a wide range of laboratory reagents and provides an ergonomic soft squeeze to help reduce hand fatigue.

Features:

  • Bottle and tubulation are molded in one piece for uniform service
  • Tip delivers a uniform, ultrafine stream without sputtering, but can also be cut back to increase flow
  • Bottle and Carton are recyclable in many communities
  • Manufactured from non-toxic materials
  • Flexible LDPE bottle provides soft, ergonomic squeeze
  • Polypropylene screw closure

Includes:
Polypropylene screw closure.

Recycling information:
Nalgene reusable LDPE wash bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Nalgene LDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene LDPE bottles.

Specifications
Capacity16 oz., 500 mL
Closure MaterialPolypropylene
Closure Size38 mm
Closure TypeScrew Cap
DescriptionLDPE bottle/tubulation
Diameter (Metric)76.7 mm
Height (English)6.81 in.
Height (Metric)173 mm
No. per Case24
No. per Pack4
Through the Closure DispensingNo
Capacity (English)16 oz.
Capacity (Metric)500 mL
Closure IncludedYes
ColorTranslucent
GraduatedNo
MaterialLDPE
MouthWide
Product TypeWash Bottle
ShapeCylindrical
Unit SizePack of 4