KingFisher Apex Accessories
Actual product may vary
KingFisher Apex Accessories
KingFisher Apex Accessories
KingFisher Apex Accessories
KingFisher Apex Accessories
KingFisher Apex Accessories
KingFisher Apex Accessories
Thermo Scientific™

KingFisher Apex Accessories

KingFisher Apex is a flexible system for different types of sample processing needs which offers highly versatile, automated magnetic-particle processing for DNA/RNA, protein and cell purification from virtually any source.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct Type
2407991096 PCR Head
24075970Matrix Heating Block
24079920Combi Head
2407993096 DW Head
2407994024 Combi Head
24075910PCR Plate Heating Block
2407592096 KF Heating Block
2407593096 DW Heating Block
2407594024 DW Heating Block
24075950Heating Block 96
24075960Heating Block 24
N21445Storage Tube Adapter
N21446Plate Adapter
N21447UV Lamp (Replacement)
Catalog number 24079910
Price (INR)
711,444.00
Each
Product Type:
96 PCR Head
Price (INR)
711,444.00
Each
KingFisher Apex is a flexible system for different types of sample processing needs which offers highly versatile, automated magnetic-particle processing for DNA/RNA, protein and cell purification from virtually any source. Using revolutionary magnetic particle separation technology, this system provides excellent reproducibility and quality
  • User-friendly high throughput sample purification system for automating DNA/RNA, protein and cell purification
  • Provides versatile and high-speed purification with reproducible results
  • Easy to use interface with touch screen empowering the user to control all details of the application and purification process
  • BindIx PC Software for added convenience
  • Extensive selection of validated protocols for different type of applications
  • Cloud library for quick protocol retrieval
Specifications
For Use With (Equipment)KingFisher Apex
Magnetic Rods96
Particle Size (Metric)ca. >1 μm
Plate Type96 PCR plate (semi/skirted)
Type of Magnet Head96 PCR
Capacity96 Samples
Format96-well Plate
Product Type96 PCR Head
Volume (Metric) Processing10 to 80 μL
Unit SizeEach