Silica gel, for chromatography, 0.060-0.200 mm, 40 A
Silica gel, for chromatography, 0.060-0.200 mm, 40 A
Silica gel, for chromatography, 0.060-0.200 mm, 40 A
Silica gel, for chromatography, 0.060-0.200 mm, 40 A
Acros Organics

Silica gel, for chromatography, 0.060-0.200 mm, 40 A

CAS: 7631-86-9 | O2Si | 60.08 g/mol
Have Questions?
Catalog NumberQuantity
2416600101 kg
Catalog number 241660010
HSN 28112200
Price (INR)
16,807.00
Each
Quantity:
1 kg
Request bulk or custom format
Price (INR)
16,807.00
Each
Chemical Identifiers
CAS7631-86-9
IUPAC Namesilanedione
Molecular FormulaO2Si
InChI KeyVYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILESO=[Si]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Residual water2 to 4 % (160°C)
Particle size=<10 % (Undersize)
Appearance (Form)Powder
pH5.5 to 8.0 (5 % suspension in water)
Particle size0.060 to 0.200 mm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For column chromatography

RUO – Research Use Only