3,4-Difluoronitrobenzene, 99%
3,4-Difluoronitrobenzene, 99%
3,4-Difluoronitrobenzene, 99%
3,4-Difluoronitrobenzene, 99%
Acros Organics

3,4-Difluoronitrobenzene, 99%

CAS: 369-34-6 | C6H3F2NO2 | 159.09 g/mol
Have Questions?
Catalog NumberQuantityPackaging
241930250
also known as 24193
25 mLGlass bottle
Catalog number 241930250
also known as 24193
HSN 29049090
Price (INR)
24,358.00
Each
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
24,358.00
Each
Add to cart
Chemical Identifiers
CAS369-34-6
IUPAC Name1,2-difluoro-4-nitrobenzene
Molecular FormulaC6H3F2NO2
InChI KeyRUBQQRMAWLSCCJ-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(F)C(F)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.5080 to 1.5100 (20°C, 589 nm)
Appearance (Color)Clear yellow
Appearance (Form)Liquid
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only