Nalgene™ Color-Coded LDPE Wash Bottles
Actual product may vary
Nalgene™ Color-Coded LDPE Wash Bottles
Nalgene™ Color-Coded LDPE Wash Bottles
Nalgene™ Color-Coded LDPE Wash Bottles

Nalgene™ Color-Coded LDPE Wash Bottles

Simplify content identification with these narrow-mouth wash bottles that feature colored closures with an integrated stem.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberClosure Color
2422-2500Yellow
2422-3500Blue
2422-5500Red
2422-4500Green
Catalog number 2422-2500
HSN 39233090
Price (INR)
29,928.00
Case
Closure Color:
Yellow
Price (INR)
29,928.00
Case

High quality squeeze bottles with color-coded closures for easy identification. Thermo Scientific™ Nalgene™ Color-Coded LDPE Wash Bottles are made from lab quality low density polyethylene (LDPE) for dependably low leachables and chemical compatability with a wide range of lab reagents. Enjoy smooth, precision, sputter-free fluid dispensing with an ergonomic soft squeeze. Ideal for rinsing tubes and lab apparatus, bringing volumetric flasks to final volume and other precision rinsing and solution dispensing needs. The dispensing nozzle is molded in one piece with the bottle closure eliminating risk of separation when the bottle is squeezed. Cut back tip to increase flow.

  • Color-coded top allows you to coordinate reagents with industry storage codes.
  • Tip can be cut back to provide increased flow.
  • Tight fit of draw tube and closure prevents separation when squeezing bottle.
  • Flexible LDPE bottle provides soft, ergonomic squeeze.
  • Sputter-free, precision stream.
  • Bottle and Carton are readily recyclable in most communities.
  • Manufactured from non-toxic materials.

Recycling Information:
Nalgene reusable LDPE wash bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Nalgene LDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene LDPE bottles.

Specifications
Capacity (Metric)500 mL
Capacity (English)16 oz.
ColorNatural
DescriptionWith stem and draw tube
MaterialLDPE
No. per Case24
No. per Pack6
Closure ColorYellow
Closure IncludedYes
GraduatedNo
MouthNarrow
ShapeCylindrical
Height (Metric)224 mm
Product TypeColor-coded Wash Bottle
Height (English)8.8 in.
Closure MaterialPolypropylene
Closure Size28 mm
Diameter (Metric)72.9 mm
Closure TypeScrew Cap
Through the Closure DispensingYes
AutoclavableNot Autoclavable
Unit SizeCase