Nunc™ 384-Well Optical-Bottom Microplate, blac, non-treated surface, no lid, non-sterile, Pack of 10
Thermo Scientific™
Nunc™ 384-Well Optical-Bottom Microplate, blac, non-treated surface, no lid, non-sterile, Pack of 10
Achieve optimal fluorescence and luminescence in cell culture and microscopic applications with Nunc MicroWell 384-well optical-bottom plates, featuring a polystyrene black upper structure with a polystyrene film at the bottom of the plate.
Have Questions?
Catalog NumberQuantity
24276430/Cs.
Catalog number 242764
Price (INR)
36,302.00
Case of 30
Quantity:
30/Cs.
Thermo Scientific Nunc MicroWell 384-well optical-bottom plates with polymer base have a solid polystyrene black upper structure with a polystyrene film at the bottom of the plate. The plates are optimized for fluorescence and luminescence in cell culture and microscopic applications.
Features
  • Cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth
  • Untreated polystyrene is hydrophobic and suited for growth of suspension cultures that can proliferate and grow without attachment
  • Polystyrene upper structure is available with either a #1.5 cover glass or polystyrene base
  • Customized bar coding is available
Color: black with clear bottom
Sterility: non-sterile
Lid: without lid
Type: optical polymer base
Specifications
Certifications/ComplianceAll microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
LidWithout Lid
No. per Case30
No. per Pack10
Packaging10 Pk., 30 Cases
Product TypeOptical Polymer Base
Quantity30/Cs.
Array16 x 24
BarcodeCustom Barcoding Available
ColorBlack
MaterialPolystyrene
No. of Wells384
Product LineNunc
SterilityNon-sterile
Surface TreatmentNon-treated
Well ShapeFlat Bottom (F-well)
Unit SizeCase of 30