UltraPure™ DNA Typing Grade™ 50X TAE Buffer
Invitrogen™

UltraPure™ DNA Typing Grade™ 50X TAE Buffer

UltraPure™ DNA Typing Grade™ 50X TAE Buffer is a sterile-filtered solution of 2 M Tris-acetate and 50 mM EDTA. TAERead more
Have Questions?
Catalog NumberQuantity
247100301 L
Catalog number 24710030
Price (INR)
11,099.00
Online offer
Ends: 31-Dec-2026
18,498.00
Save 7,399.00 (40%)
Each
Quantity:
1 L
Request bulk or custom format
Price (INR)
11,099.00
Online offer
Ends: 31-Dec-2026
18,498.00
Save 7,399.00 (40%)
Each
UltraPure™ DNA Typing Grade™ 50X TAE Buffer is a sterile-filtered solution of 2 M Tris-acetate and 50 mM EDTA. TAE Buffer is the most commonly used buffer for agarose DNA electrophoresis. To prepare a 1X solution, mix one volume of UltraPure™ 50X TAE Buffer with 49 volumes of distilled water. A 1X TAE Buffer solution contains 40 mM Tris-acetate and 1 mM EDTA with a pH of 8.3 at 23“C.

Performance and Quality Testing: No DNase activity detected.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
BufferRunning Buffer
Concentration50 X
For Use With (Application)Nucleic Acid Gel Electrophoresis, Blotting
Gel CompatibilityE-Gel™, Agarose Gels
Product LineUltraPure
Product TypeTAE Buffer
Quantity1 L
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
Store at room temperature.