meso-1,2-Diphenyl-1,2-ethanediol, 95%
meso-1,2-Diphenyl-1,2-ethanediol, 95%
meso-1,2-Diphenyl-1,2-ethanediol, 95%
Acros Organics

meso-1,2-Diphenyl-1,2-ethanediol, 95%

CAS: 579-43-1 | C14H14O2 | 214.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Packaging:
Glass bottle
Catalog number 247680050
also known as 24768
HSN 29062990
Price (INR)
12,996.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
12,996.00
Each
Chemical Identifiers
CAS579-43-1
IUPAC Name1,2-diphenylethane-1,2-diol
Molecular FormulaC14H14O2
InChI KeyIHPDTPWNFBQHEB-UHFFFAOYNA-N
SMILESOC(C(O)C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
GC>=94 %
Appearance (Form)Crystalline powder and or lumps
Melting point134°C to 140°C
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only