Nunc™ Microplate Lids
Actual product may vary
Nunc™ Microplate Lids
Nunc™ Microplate Lids
Nunc™ Microplate Lids
Nunc™ Microplate Lids

Nunc™ Microplate Lids

Protect samples from contamination and evaporation during assay processing, incubation or storage with these microplate lids.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPlate CompatibilitySterilitydimCondensationRingsNo. per Pack
250002All MicroplatesSterile20
250003All MicroplatesNon-sterile20
250005All MicroplatesNon-sterile20
253623All MicroplatesNon-sterile25
253624All MicroplatesSterile25
26333996-well PlateNon-sterile5
26412296-well PlateSterile1
264611384-well PlateSterile1
264612384-well PlateNon-sterile20
264616384-well PlateSterile20
26462396-well PlateSterile20
448143384/1536-well Plate, OmniTrayNon-sterile20
Catalog number 250002
HSN 39235090
Price (INR)
23,531.00
CS
Plate Compatibility:
All Microplates
Sterility:
Sterile
No. per Pack:
20
Price (INR)
23,531.00
CS
Protect samples from contamination and evaporation during assay processing, incubation or storage with Thermo ScientificTM NuncTM Microplate Lids.
  • Lids are for use with any standard plates
  • 96 Well Lids have individual condensation rings
  • Lids have notches for quick orientation
  • Universal Lids can be used on any standard height plate on the market
Specifications
Alternate Part NumberPIC 200-DIA 200-B7541 (BRANDED)
ColorClear
Dimensions (L x W x H)5 x 3.35 x 0.33 in. (127 x 85 x 8.34 mm)
MaterialPolystyrene
SterilitySterile
No. of Wells384/1536
Height (Metric)8.34 mm
Length (English)5 in.
Length (Metric)127 mm
Condensation RingsNo
Height (English)0.33 in.
No. per Case60
Diameter (Metric)200 mm
No. per Pack20
Dimensions (L x W)5 x 3.35 in. (127 x 85 mm)
Evaporation BarrierNo
Width (English)3.35 in.
Width (Metric)85 mm
Product TypeUniversal Lid
Product LineNunc
AdhesiveNone
ShapeRound
Temperature Range (Metric)-20°C to +80°C
Well ShapeAll well shapes
Plate CompatibilityAll Microplates
Cut-off CornersNo
Unit SizeCS