BiTek™ Tryptone
Gibco™

BiTek™ Tryptone

BiTek Tryptone is a pancreatic digest of casein. It was developed while investigating a peptone particularly suitable for the elaborationRead more
Have Questions?
Catalog NumberQuantity
25142010 kg
Catalog number 251420
Price (INR)
204,736.00
Each
Quantity:
10 kg
Price (INR)
204,736.00
Each
BiTek Tryptone is a pancreatic digest of casein. It was developed while investigating a peptone particularly suitable for the elaboration of indole by bacteria. The final product goes through fewer refinement steps during processing than Bacto Tryptone.

Specifications
For Use With (Application)Microbial protein and viral vaccine production
Product LineBiTek
Quantity10 kg
SourceAnimal Origin, Casein
FormPowder
Product TypeDehydrated Culture Media
Unit SizeEach
Contents & Storage
Storage conditions: 2°C to 25°C.  Store in cool, dry conditions in well-sealed containers.

Frequently asked questions (FAQs)

What alternative product do you recommend for Bacto Tryptone (Cat. No. 211699, 211701, 211705)?

We have several casein digests that could serve as alternatives to Bactone Tryptone (Cat. No. 211699, 211791, 211705), including Bacto Casitone (Cat. No. 225930, 225910), Difco Tryptone Peptone (Cat. No. 21921, 211922, 211923), BiTek Tryptone (Cat. No. 251420). However, the digestion and nutritive profile of each peptone are different, therefore there may be differences in bacterial growth or supplementation ability. We recommend empirically determining the performance of alternative casein digestion in your application.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.